Assay Details
| Gene Symbol: Gk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079916.3 | NP_524655.1 | 2 - 3 | 1612 | 81 | |
| NM_167803.2 | NP_728486.1 | 3 - 4 | 1385 | 81 | ||
| GenBank mRNA | AY051456.1 | - | 2 - 3 | 1462 | 81 | |
More Information
Set Membership: |
|

Gk1
Genomic Map