Assay Details
| Gene Symbol: sbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079921.3 | NP_524660.1 | 2 - 3 | 756 | 84 | |
| GenBank mRNA | AJ251947.2 | - | 2 - 3 | 720 | 84 | |
| AJ318090.1 | - | 2 - 3 | 755 | 84 | ||
More Information
Set Membership: |
|

sbr
Genomic Map