Assay Details
| Gene Symbol: sbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369981.1 | NP_001356941.1 | 6 - 7 | 1096 | 67 | |
| NM_001369982.1 | NP_001356942.1 | 5 - 6 | 1333 | 67 | ||
| NM_079921.3 | NP_524660.1 | 8 - 9 | 1923 | 67 | ||
| GenBank mRNA | AJ251947.2 | - | 8 - 9 | 1887 | 67 | |
| AJ318090.1 | - | 8 - 9 | 1922 | 67 | ||
More Information
Set Membership: |
|

sbr
Genomic Map