Assay Details
| Gene Symbol: sls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259622.1 | NP_001246551.1 | 11 - 12 | 12828 | 63 | |
| NM_001274375.1 | NP_001261304.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274376.1 | NP_001261305.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274377.1 | NP_001261306.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274378.1 | NP_001261307.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274379.1 | NP_001261308.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274380.1 | NP_001261309.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274381.1 | NP_001261310.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274382.1 | NP_001261311.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274383.1 | NP_001261312.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274384.1 | NP_001261313.1 | 10 - 11 | 12783 | 63 | ||
| NM_001274385.1 | NP_001261314.1 | 10 - 11 | 12783 | 63 | ||
| NM_079937.4 | NP_524676.2 | 10 - 11 | 12783 | 63 | ||
More Information
Set Membership: |
|

sls
Genomic Map