Assay Details
| Gene Symbol: Mcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079949.3 | NP_524688.1 | 6 - 7 | 5248 | 63 | |
| GenBank mRNA | AY058526.1 | - | 6 - 7 | 5214 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcr| Gene Symbol: Mcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079949.3 | NP_524688.1 | 6 - 7 | 5248 | 63 | |
| GenBank mRNA | AY058526.1 | - | 6 - 7 | 5214 | 63 | |
Set Membership: |
|