Assay Details
| Gene Symbol: Mgstl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079957.4 | NP_524696.2 | 1 - 2 | 239 | 80 | |
| GenBank mRNA | AY075452.1 | - | 1 - 2 | 237 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgstl| Gene Symbol: Mgstl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079957.4 | NP_524696.2 | 1 - 2 | 239 | 80 | |
| GenBank mRNA | AY075452.1 | - | 1 - 2 | 237 | 80 | |
Set Membership: |
|