Assay Details
| Gene Symbol: Nox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103866.2 | NP_001097336.1 | 5 - 6 | 1643 | 84 | |
| NM_001299556.1 | NP_001286485.1 | 5 - 6 | 1747 | 84 | ||
| NM_001299557.1 | NP_001286486.1 | 5 - 6 | 3523 | 84 | ||
| NM_137272.2 | NP_611116.2 | 5 - 6 | 1850 | 84 | ||
More Information
Set Membership: |
|

Nox
Genomic Map