Assay Details
| Gene Symbol: S-Lap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137288.3 | NP_611132.1 | 1 - 2 | 315 | 72 | |
| GenBank mRNA | AY047564.1 | - | 1 - 2 | 273 | 72 | |
More Information
Set Membership: |
|

S-Lap8
Genomic Map