Assay Details
| Gene Symbol: CG8303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299566.1 | NP_001286495.1 | 4 - 5 | 1393 | 90 | |
| NM_137297.3 | NP_611141.3 | 4 - 5 | 1417 | 90 | ||
More Information
Set Membership: |
|

CG8303
Genomic Map