Assay Details
| Gene Symbol: unc-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103876.3 | NP_001097346.1 | 11 - 12 | 1770 | 69 | |
| NM_001259444.2 | NP_001246373.1 | 11 - 12 | 1770 | 69 | ||
| NM_001259445.2 | NP_001246374.1 | 11 - 12 | 1770 | 69 | ||
| NM_001274104.2 | NP_001261033.1 | 9 - 10 | 1570 | 69 | ||
| NM_001316424.1 | NP_001303353.1 | 9 - 10 | 1570 | 69 | ||
| NM_137311.5 | NP_611155.3 | 10 - 11 | 1728 | 69 | ||
| NM_166190.4 | NP_725607.2 | 10 - 11 | 1728 | 69 | ||
| NM_166192.4 | NP_725610.2 | 9 - 10 | 1737 | 69 | ||
More Information
Set Membership: |
|

unc-104
Genomic Map