Assay Details
| Gene Symbol: unc-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103876.3 | NP_001097346.1 | 20 - 21 | 4442 | 66 | |
| NM_001259444.2 | NP_001246373.1 | 20 - 21 | 4442 | 66 | ||
| NM_001274104.2 | NP_001261033.1 | 18 - 19 | 4242 | 66 | ||
| NM_001316424.1 | NP_001303353.1 | 18 - 19 | 4242 | 66 | ||
| NM_137311.5 | NP_611155.3 | 20 - 21 | 4607 | 66 | ||
| NM_166190.4 | NP_725607.2 | 19 - 20 | 4400 | 66 | ||
| NM_166192.4 | NP_725610.2 | 18 - 19 | 4409 | 66 | ||
More Information
Set Membership: |
|

unc-104
Genomic Map