Assay Details
| Gene Symbol: unc-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103876.3 | NP_001097346.1 | 21 - 22 | 4837 | 72 | |
| NM_001259444.2 | NP_001246373.1 | 21 - 22 | 4837 | 72 | ||
| NM_001259445.2 | NP_001246374.1 | 21 - 22 | 4828 | 72 | ||
| NM_001274104.2 | NP_001261033.1 | 19 - 20 | 4637 | 72 | ||
| NM_001316424.1 | NP_001303353.1 | 19 - 20 | 4637 | 72 | ||
| NM_137311.5 | NP_611155.3 | 21 - 22 | 5002 | 72 | ||
| NM_166190.4 | NP_725607.2 | 20 - 21 | 4795 | 72 | ||
| NM_166192.4 | NP_725610.2 | 19 - 20 | 4804 | 72 | ||
More Information
Set Membership: |
|

unc-104
Genomic Map