Assay Details
| Gene Symbol: unc-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103876.3 | NP_001097346.1 | 23 - 24 | 4979 | 69 | |
| NM_001259445.2 | NP_001246374.1 | 23 - 24 | 4970 | 69 | ||
| NM_137311.5 | NP_611155.3 | 23 - 24 | 5144 | 69 | ||
| NM_166190.4 | NP_725607.2 | 22 - 23 | 4937 | 69 | ||
| NM_166192.4 | NP_725610.2 | 21 - 22 | 4946 | 69 | ||
More Information
Set Membership: |
|

unc-104
Genomic Map