Assay Details
| Gene Symbol: AsnRS-m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137332.3 | NP_611176.1 | 1 - 2 | 126 | 89 | |
| GenBank mRNA | AY071089.1 | - | 1 - 2 | 106 | 89 | |
More Information
Set Membership: |
|

AsnRS-m
Genomic Map