Assay Details
| Gene Symbol: Cda9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299590.1 | NP_001286519.1 | 1 - 2 | 569 | 68 | |
| NM_137348.4 | NP_611192.1 | 1 - 2 | 238 | 68 | ||
| GenBank mRNA | BT088821.1 | - | 1 - 2 | 569 | 68 | |
More Information
Set Membership: |
|

Cda9
Genomic Map