Assay Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299592.1 | NP_001286521.1 | 1 - 2 | 177 | 82 | |
| NM_137351.3 | NP_611195.1 | 1 - 2 | 177 | 82 | ||
| GenBank mRNA | AY051489.1 | - | 1 - 2 | 161 | 82 | |
More Information
Set Membership: |
|

Gbp2
Genomic Map