Assay Details
| Gene Symbol: Camp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299594.1 | NP_001286523.1 | 2 - 3 | 1041 | 72 | |
| NM_137360.2 | NP_611204.2 | 3 - 4 | 1133 | 72 | ||
| GenBank mRNA | AY121689.1 | - | 3 - 4 | 1131 | 72 | |
More Information
Set Membership: |
|

Camp
Genomic Map