Assay Details
| Gene Symbol: CG6550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137363.4 | NP_611207.1 | 1 - 2 | 1072 | 64 | |
| GenBank mRNA | AY051634.1 | - | 1 - 2 | 1040 | 64 | |
More Information
Set Membership: |
|

CG6550
Genomic Map