Assay Details
| Gene Symbol: CG4984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144223.3 | NP_001137695.1 | 2 - 3 | 1117 | 65 | |
| NM_001169712.2 | NP_001163183.1 | 2 - 3 | 1117 | 65 | ||
| GenBank mRNA | BT044241.1 | - | 1 - 2 | 1113 | 65 | |
| Gene Symbol: CG4975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144221.3 | NP_001137693.2 | 2 - 3 | 1117 | 65 | |
| NM_001144222.2 | NP_001137694.1 | 2 - 3 | 1117 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT044241.1, NM_001144223.3, NM_001169712.2 transcripts.
Set Membership: |
|

CG4984
Genomic Map