Assay Details
| Gene Symbol: CG4984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144223.3 | NP_001137695.1 | 4 - 5 | 1766 | 70 | |
| NM_001169711.2 | NP_001163182.1 | 2 - 3 | 623 | 70 | ||
| NM_001169712.2 | NP_001163183.1 | 4 - 5 | 1766 | 70 | ||
| NM_137410.4 | NP_611254.2 | 2 - 3 | 623 | 70 | ||
| GenBank mRNA | BT044241.1 | - | 3 - 4 | 1762 | 70 | |
| BT133275.1 | - | 2 - 3 | 598 | 70 | ||
| Gene Symbol: CG4975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144221.3 | NP_001137693.2 | 4 - 5 | 1766 | 70 | |
More Information
Set Membership: |
|

CG4984
Genomic Map