Assay Details
| Gene Symbol: CG4984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144223.3 | NP_001137695.1 | 6 - 7 | 2528 | 71 | |
| NM_137410.4 | NP_611254.2 | 4 - 5 | 1385 | 71 | ||
| GenBank mRNA | BT044241.1 | - | 5 - 6 | 2524 | 71 | |
| Gene Symbol: CG4975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144221.3 | NP_001137693.2 | 6 - 7 | 2528 | 71 | |
More Information
Set Membership: |
|

CG4984
Genomic Map