Assay Details
| Gene Symbol: PIG-O | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137488.3 | NP_611332.1 | 2 - 3 | 1106 | 67 | |
| GenBank mRNA | BT150350.1 | - | 2 - 3 | 1063 | 67 | |
More Information
Set Membership: |
|

PIG-O
Genomic Map