Assay Details
| Gene Symbol: Rgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299648.1 | NP_001286577.1 | 2 - 3 | 470 | 60 | |
| NM_137498.3 | NP_611342.1 | 2 - 3 | 857 | 60 | ||
| GenBank mRNA | BT011477.1 | - | 2 - 3 | 857 | 60 | |
More Information
Set Membership: |
|

Rgk2
Genomic Map