Assay Details
| Gene Symbol: Rgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274158.1 | NP_001261087.1 | 1 - 2 | 300 | 70 | |
| NM_137567.3 | NP_611411.2 | 1 - 2 | 300 | 70 | ||
| GenBank mRNA | BT010242.1 | - | 1 - 2 | 294 | 70 | |
More Information
Set Membership: |
|

Rgk1
Genomic Map