Assay Details
| Gene Symbol: Rgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274158.1 | NP_001261087.1 | 3 - 4 | 700 | 69 | |
| NM_137567.3 | NP_611411.2 | 3 - 4 | 700 | 69 | ||
| NM_166341.3 | NP_725875.3 | 10 - 11 | 4197 | 69 | ||
| GenBank mRNA | BT010242.1 | - | 3 - 4 | 694 | 69 | |
More Information
Set Membership: |
|

Rgk1
Genomic Map