Assay Details
| Gene Symbol: Rgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137567.3 | NP_611411.2 | 5 - 6 | 1061 | 84 | |
| NM_166341.3 | NP_725875.3 | 12 - 13 | 4558 | 84 | ||
| GenBank mRNA | BT010242.1 | - | 5 - 6 | 1055 | 84 | |
More Information
Set Membership: |
|

Rgk1
Genomic Map