Assay Details
| Gene Symbol: Rgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274158.1 | NP_001261087.1 | 7 - 8 | 1199 | 72 | |
| NM_137567.3 | NP_611411.2 | 6 - 7 | 1151 | 72 | ||
| NM_166341.3 | NP_725875.3 | 13 - 14 | 4648 | 72 | ||
| GenBank mRNA | BT010242.1 | - | 6 - 7 | 1145 | 72 | |
More Information
Set Membership: |
|

Rgk1
Genomic Map