Assay Details
| Gene Symbol: Rgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274158.1 | NP_001261087.1 | 9 - 10 | 1531 | 79 | |
| NM_137567.3 | NP_611411.2 | 8 - 9 | 1483 | 79 | ||
| NM_166341.3 | NP_725875.3 | 15 - 16 | 4980 | 79 | ||
| GenBank mRNA | BT010242.1 | - | 8 - 9 | 1477 | 79 | |
More Information
Set Membership: |
|

Rgk1
Genomic Map