Assay Details
| Gene Symbol: Obp56e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137601.4 | NP_611445.1 | 1 - 2 | 105 | 61 | |
| GenBank mRNA | AY070654.1 | - | 1 - 2 | 103 | 61 | |
| KX531903.1 | - | 1 - 2 | 53 | 61 | ||
More Information
Set Membership: |
|

Obp56e
Genomic Map