Assay Details
| Gene Symbol: Hmg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137709.4 | NP_611553.1 | 1 - 2 | 500 | 104 | |
| GenBank mRNA | AY058722.1 | - | 1 - 2 | 484 | 104 | |
More Information
Set Membership: |
|

Hmg-2
Genomic Map