Assay Details
| Gene Symbol: Rgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032275.2 | NP_001027446.2 | 7 - 8 | 2464 | 67 | |
| NM_001103925.3 | NP_001097395.1 | 7 - 8 | 1831 | 67 | ||
| NM_001103926.4 | NP_001097396.3 | 8 - 9 | 2584 | 67 | ||
| NM_001299741.1 | NP_001286670.1 | 7 - 8 | 1831 | 67 | ||
| GenBank mRNA | KJ826443.1 | - | 6 - 7 | 1069 | 67 | |
| KJ826444.1 | - | 6 - 7 | 1069 | 67 | ||
More Information
Set Membership: |
|

Rgk3
Genomic Map