Assay Details
| Gene Symbol: Tomosyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169265.2 | NP_001162736.2 | 13 - 14 | 2671 | 99 | |
| NM_001258715.2 | NP_001245644.1 | 13 - 14 | 2671 | 99 | ||
| NM_132591.5 | NP_572819.4 | 14 - 15 | 2716 | 99 | ||
| NM_167330.3 | NP_727630.3 | 13 - 14 | 2793 | 99 | ||
More Information
Set Membership: |
|

Tomosyn
Genomic Map