Assay Details
| Gene Symbol: Pde9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272566.1 | NP_001259495.1 | 6 - 7 | 3892 | 60 | |
| NM_001272567.2 | NP_001259496.1 | 7 - 8 | 4027 | 60 | ||
| NM_132606.5 | NP_572834.2 | 7 - 8 | 4027 | 60 | ||
| NM_167340.5 | NP_727644.3 | 7 - 8 | 4027 | 60 | ||
More Information
Set Membership: |
|

Pde9
Genomic Map