Assay Details
| Gene Symbol: up | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014737.2 | NP_001014737.1 | 8 - 9 | 970 | 69 | |
| NM_001014738.3 | NP_001014738.2 | 8 - 9 | 970 | 69 | ||
| NM_001014739.2 | NP_001014739.1 | 6 - 7 | 904 | 69 | ||
| NM_001169273.2 | NP_001162744.1 | 7 - 8 | 1018 | 69 | ||
| NM_001258724.2 | NP_001245653.1 | 8 - 9 | 974 | 69 | ||
| NM_001272593.2 | NP_001259522.1 | 8 - 9 | 1045 | 69 | ||
| NM_001272594.2 | NP_001259523.1 | 8 - 9 | 970 | 69 | ||
| NM_001272595.2 | NP_001259524.1 | 7 - 8 | 946 | 69 | ||
| NM_001298284.1 | NP_001285213.1 | 7 - 8 | 994 | 69 | ||
| NM_001298285.1 | NP_001285214.1 | 7 - 8 | 1144 | 69 | ||
| NM_001298286.1 | NP_001285215.1 | 5 - 6 | 861 | 69 | ||
| NM_080349.3 | NP_525088.2 | 8 - 9 | 973 | 69 | ||
| NM_167375.2 | NP_727718.1 | 7 - 8 | 1021 | 69 | ||
| NM_167376.2 | NP_727719.1 | 5 - 6 | 1132 | 69 | ||
| GenBank mRNA | AY051989.1 | - | 5 - 6 | 1132 | 69 | |
| BT125908.1 | - | 8 - 9 | 949 | 69 | ||
| BT126178.1 | - | 7 - 8 | 940 | 69 | ||
More Information
Set Membership: |
|

up
Genomic Map