Assay Details
| Gene Symbol: be | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258745.2 | NP_001245674.1 | 1 - 2 | 526 | 82 | |
| NM_132762.4 | NP_572990.2 | 1 - 2 | 526 | 82 | ||
| NM_167424.2 | NP_727817.1 | 1 - 2 | 526 | 82 | ||
| GenBank mRNA | BT044236.1 | - | 1 - 2 | 509 | 82 | |
| BT056306.1 | - | 1 - 2 | 553 | 82 | ||
More Information
Set Membership: |
|

be
Genomic Map