Assay Details
| Gene Symbol: be | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258745.2 | NP_001245674.1 | 3 - 4 | 944 | 91 | |
| NM_132762.4 | NP_572990.2 | 3 - 4 | 944 | 91 | ||
| NM_167424.2 | NP_727817.1 | 3 - 4 | 944 | 91 | ||
| GenBank mRNA | BT044236.1 | - | 3 - 4 | 927 | 91 | |
| BT056306.1 | - | 3 - 4 | 971 | 91 | ||
More Information
Set Membership: |
|

be
Genomic Map