Assay Details
| Gene Symbol: Scamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272643.1 | NP_001259572.1 | 1 - 2 | 177 | 55 | |
| NM_132795.4 | NP_573023.2 | 1 - 2 | 177 | 55 | ||
| NM_167442.2 | NP_727850.1 | 1 - 2 | 177 | 55 | ||
| GenBank mRNA | AF241834.1 | - | 1 - 2 | 70 | 55 | |
| AY075562.1 | - | 1 - 2 | 184 | 55 | ||
More Information
Set Membership: |
|

Scamp
Genomic Map