Assay Details
| Gene Symbol: sun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167468.3 | NP_727905.1 | 2 - 3 | 252 | 91 | |
| GenBank mRNA | BT099753.1 | - | 2 - 3 | 234 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sun| Gene Symbol: sun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167468.3 | NP_727905.1 | 2 - 3 | 252 | 91 | |
| GenBank mRNA | BT099753.1 | - | 2 - 3 | 234 | 91 | |
Set Membership: |
|