Assay Details
| Gene Symbol: Traf-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258784.2 | NP_001245713.1 | 3 - 4 | 965 | 68 | |
| NM_001258785.2 | NP_001245714.1 | 3 - 4 | 1363 | 68 | ||
| NM_001272683.1 | NP_001259612.1 | 3 - 4 | 1315 | 68 | ||
| NM_001272684.1 | NP_001259613.1 | 3 - 4 | 966 | 68 | ||
| NM_132898.4 | NP_573126.1 | 2 - 3 | 1444 | 68 | ||
| NM_167517.3 | NP_727976.1 | 3 - 4 | 1373 | 68 | ||
| NM_167518.3 | NP_727977.1 | 3 - 4 | 917 | 68 | ||
| GenBank mRNA | AY052057.1 | - | 2 - 3 | 1421 | 68 | |
| BT125880.1 | - | 3 - 4 | 1333 | 68 | ||
More Information
Set Membership: |
|

Traf-like
Genomic Map