Assay Details
| Gene Symbol: AnxB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272687.1 | NP_001259616.1 | 3 - 4 | 656 | 76 | |
| NM_001272689.1 | NP_001259618.1 | 3 - 4 | 1098 | 76 | ||
| NM_167519.2 | NP_727978.1 | 2 - 3 | 643 | 76 | ||
| GenBank mRNA | BT125828.1 | - | 2 - 3 | 644 | 76 | |
More Information
Set Membership: |
|

AnxB11
Genomic Map