Assay Details
| Gene Symbol: wupA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258805.2 | NP_001245734.1 | 5 - 6 | 507 | 134 | |
| NM_001272739.1 | NP_001259668.1 | 5 - 6 | 506 | 134 | ||
| NM_001272740.2 | NP_001259669.1 | 6 - 7 | 692 | 134 | ||
| NM_001272741.1 | NP_001259670.1 | 6 - 7 | 694 | 134 | ||
| NM_167607.3 | NP_728141.1 | 5 - 6 | 507 | 134 | ||
| GenBank mRNA | BT125818.1 | - | 5 - 6 | 483 | 134 | |
More Information
Set Membership: |
|

wupA
Genomic Map