Assay Details
| Gene Symbol: Rcd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134491.4 | NP_608335.1 | 2 - 3 | 440 | 104 | |
| NM_167675.2 | NP_728284.1 | 1 - 2 | 508 | 104 | ||
| GenBank mRNA | AY118785.1 | - | 1 - 2 | 508 | 104 | |
More Information
Set Membership: |
|

Rcd-1
Genomic Map