Assay Details
| Gene Symbol: pico | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201745.1 | NP_001188674.1 | 2 - 3 | 1219 | 77 | |
| NM_001201746.1 | NP_001188675.1 | 2 - 3 | 1259 | 77 | ||
| NM_134519.3 | NP_608363.1 | 2 - 3 | 2307 | 77 | ||
| NM_167686.3 | NP_728307.1 | 2 - 3 | 1522 | 77 | ||
| GenBank mRNA | BT124745.1 | - | 2 - 3 | 1624 | 77 | |
More Information
Set Membership: |
|

pico
Genomic Map