Assay Details
| Gene Symbol: Pp4-19C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298560.1 | NP_001285489.1 | 2 - 3 | 1252 | 56 | |
| NM_001298561.1 | NP_001285490.1 | 1 - 2 | 949 | 56 | ||
| NM_001316641.1 | NP_001303570.1 | 2 - 3 | 1070 | 56 | ||
| NM_080064.3 | NP_524803.1 | 2 - 3 | 1046 | 56 | ||
| NM_167703.4 | NP_728342.1 | 2 - 3 | 1094 | 56 | ||
| GenBank mRNA | AY113503.1 | - | 2 - 3 | 1268 | 56 | |
More Information
Set Membership: |
|

Pp4-19C
Genomic Map