Assay Details
| Gene Symbol: GstT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169337.2 | NP_001162808.1 | 2 - 3 | 279 | 74 | |
| NM_001298566.1 | NP_001285495.1 | 3 - 4 | 670 | 74 | ||
| NM_167706.3 | NP_728347.1 | 2 - 3 | 176 | 74 | ||
More Information
Set Membership: |
|

GstT3
Genomic Map