Assay Details
| Gene Symbol: tty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272837.2 | NP_001259766.1 | 3 - 4 | 860 | 57 | |
| NM_001272838.1 | NP_001259767.1 | 3 - 4 | 1021 | 57 | ||
| NM_001272839.2 | NP_001259768.1 | 3 - 4 | 959 | 57 | ||
| NM_001272840.2 | NP_001259769.1 | 3 - 4 | 1019 | 57 | ||
| NM_080712.7 | NP_542443.4 | 3 - 4 | 1021 | 57 | ||
| NM_167744.4 | NP_728415.4 | 3 - 4 | 1619 | 57 | ||
| GenBank mRNA | AF184227.1 | - | 3 - 4 | 938 | 57 | |
More Information
Set Membership: |
|

tty
Genomic Map