Assay Details
| Gene Symbol: Cda4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167788.2 | NP_728468.1 | 5 - 6 | 673 | 99 | |
| GenBank mRNA | AY113493.1 | - | 5 - 6 | 672 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cda4| Gene Symbol: Cda4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167788.2 | NP_728468.1 | 5 - 6 | 673 | 99 | |
| GenBank mRNA | AY113493.1 | - | 5 - 6 | 672 | 99 | |
Set Membership: |
|