Assay Details
| Gene Symbol: Cda4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167788.2 | NP_728468.1 | 7 - 8 | 1280 | 67 | |
| GenBank mRNA | AY113493.1 | - | 7 - 8 | 1279 | 67 | |
More Information
Set Membership: |
|

Cda4
Genomic Map