Assay Details
| Gene Symbol: E(bx) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169833.2 | NP_001163304.1 | 12 - 13 | 8389 | 147 | |
| NM_001274258.1 | NP_001261187.1 | 13 - 14 | 8066 | 147 | ||
| NM_001274259.1 | NP_001261188.1 | 13 - 14 | 8710 | 147 | ||
| NM_001274260.1 | NP_001261189.1 | 12 - 13 | 8019 | 147 | ||
| NM_001274261.1 | NP_001261190.1 | 12 - 13 | 7931 | 147 | ||
| NM_167817.3 | NP_728505.1 | 13 - 14 | 8374 | 147 | ||
| NM_167819.3 | NP_728507.1 | 12 - 13 | 8434 | 147 | ||
More Information
Set Membership: |
|

E(bx)
Genomic Map