Assay Details
| Gene Symbol: Atac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144394.2 | NP_001137866.1 | 4 - 5 | 905 | 96 | |
| NM_001259602.2 | NP_001246531.1 | 6 - 7 | 1028 | 96 | ||
| NM_001274275.1 | NP_001261204.1 | 5 - 6 | 926 | 96 | ||
| NM_167828.4 | NP_728520.2 | 5 - 6 | 1058 | 96 | ||
More Information
Set Membership: |
|

Atac3
Genomic Map